Key Documents
BAF203201660
SP Bel-Art® Safe-Lab Joint Stirrer Bearing
for 24/40 Tapered Joints
Select a Size
1 EACH
€183.00
€183.00
Please contact Customer Service for Availability
Select a Size
Change View
1 EACH
€183.00
About This Item
UNSPSC Code:
41122800
NACRES:
NB.43
€183.00
Please contact Customer Service for Availability
packaging
pack of 1 ea
manufacturer/tradename
Bel-Art F20320-1660
General description
For use with 10mm stirring rods, the body is PTFE with a Viton® O-ring.
Features and Benefits
  • Comes complete with two PTFE inserts with inside O-ring for fine adjustment of stirring rod tension
  • Threaded polypropylene locking compression cap on top
  • Extracting nut above the ground joint allows easy and quick removal from flask
  • Not autoclavable
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Viton is a registered trademark of E. I. du Pont de Nemours and Company
Choose from one of the most recent versions:
Certificates of Analysis (COA)