Merck
All Photos(6)
Key Documents
BAB609100000
SP Bel-Art® Durac® Bluetooth Thermometer
temp. range at -50-300 °C, stainless steel stem(s)
Synonym(s):
Bluetooth Stainless Steel Stem Thermometer, Bluetooth Temperature Sensor
About This Item
UNSPSC Code:
41112303
NACRES:
NB.79
material
stainless steel stem(s)
packaging
pack of 1 ea
manufacturer/tradename
HB Instruments B60910-0000
range of application measuring range
-50-300 °C, resolution: 1 °F/C (±1°C from -20/130°C; ±2°C from -50/-21°C and 131/200°C; otherwise ±4°C)
parameter
temp. range at -50-300 °C
probe L
145 mm , 5.8"
battery power
(CR2032)
General description
Remotely monitor temperature by broadcasting readings to any Apple or Android smartphone or device with Bluetooth 4.0 or better utilizing the free THERMSmart app.
Features and Benefits
  • Monitor critical environments without a physical connection; maximum signal distance is approximately 40 meters
  • Alarm sounds to alert user if temperature falls outside of the set parameters Includes countdown timer function
  • Alarm and timer settings savable for reuse
  • LCD screen for on-unit monitoring
  • App can trigger indicator light for unit identification -Indicators include active alarm state and low battery
  • Verification model provides accurate temperature reading without sample contamination by using a glass bead filled bottle (included) which mimics the sample
  • Includes Certificate of Compliance indicating accuracy for 4 temperature points and multi-language (English, French, German, Italian, Portuguese and Spanish) instructions
  • Individually serialized, CE marked and RoHS compliant -Batteries included
Legal Information
Bel-Art is a registered trademark of SP Scienceware, a division of SP
Durac is a registered trademark of SP Scienceware, a division of SP
Choose from one of the most recent versions:
Certificates of Analysis (COA)