Skip to Content
Merck
All Photos(1)

Key Documents

A76427

Sigma-Aldrich

3-Aminopropionitrile fumarate salt

≥99%

Synonym(s):

β-Alaninenitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH2CH2CH2CN · ½ (HO2CCH=CHCO2H)
CAS Number:
Molecular Weight:
128.13
Beilstein:
3573532
EC Number:
UNSPSC Code:
12352103

Assay

≥99%

mp

177 °C (dec.) (lit.)

SMILES string

NCCC#N.NCCC#N.OC(=O)\C=C\C(O)=O

InChI

1S/C4H4O4.2C3H6N2/c5-3(6)1-2-4(7)8;2*4-2-1-3-5/h1-2H,(H,5,6)(H,7,8);2*1-2,4H2/b2-1+;;

InChI key

NYMXYZMHOZAPHQ-SEPHDYHBSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

In mammals, metabolism of 3-aminopropionitrile leads to release of cyanide ions. Metabolic activity is localized in the microsomal fraction of the liver, and is believed to proceed through a free-radical pathway.

replaced by

Product No.
Description
Pricing

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Repr. 1B

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

G Granström et al.
Scandinavian journal of plastic and reconstructive surgery and hand surgery, 26(1), 13-20 (1992-01-01)
Cleft palate was induced in 420 embryos of Sprague-Dawley rats with a single oral dose of 600 mg/kg beta-aminoproprionitrile (BAPN) on embryonal day 15, 7 hours. The cleft palate was accompanied by a pathological differentiation pattern of various isoenzymes in
D L Hoffman et al.
Experimental and molecular pathology, 39(2), 154-162 (1983-10-01)
beta-Aminopropionitrile as free base (BAPN) was applied onto the incised or intact skin of rats at the dose of 5, 20, 100, and 200 microliters for 9 days, twice daily. Breaking strength of the skin wound or intact skin was
Hirotaka Nagashima et al.
Journal of vascular surgery, 36(4), 818-823 (2002-10-09)
Cystic medial degeneration (CMD) is a histologic abnormality that is common in aortic diseases such as aortic dilation, aneurysm, or dissection. Although little is known about the mechanism underlying CMD, we have previously demonstrated that angiotensin II signaling via angiotensin
S Gunasekaran et al.
Surgical neurology, 28(3), 201-207 (1987-09-01)
The purpose of this study is to evaluate possible toxic effects of beta-aminopropionitrile fumarate (beta APN), a lathyrogenic agent that inhibits fibrosis. This drug has been considered for use as an adjunct to surgical repair after topical application upon peripheral
L Feuer et al.
Endokrinologie, 75(3), 350-356 (1980-06-01)
BAPN-induced osteolathyrism in young and older male rats was examined for its responses to parathormone-free, gel-filtered bovine parathyroid extract (extract P) and to glutaurine (gamma-L-glutamyl-taurine), the latter being a hitherto unknown putative hormone isolated from bovine parathyroid and reproduced by

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service