Skip to Content
Merck
All Photos(1)

Key Documents

A70801

Sigma-Aldrich

2-Amino-5-nitropyridine

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H5N3O2
CAS Number:
Molecular Weight:
139.11
Beilstein:
120353
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

mp

186-188 °C (lit.)

SMILES string

Nc1ccc(cn1)[N+]([O-])=O

InChI

1S/C5H5N3O2/c6-5-2-1-4(3-7-5)8(9)10/h1-3H,(H2,6,7)

InChI key

UGSBCCAHDVCHGI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

435.2 °F

Flash Point(C)

224 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Danila V Zimenkov et al.
Journal of clinical microbiology, 53(4), 1103-1114 (2015-01-23)
In addition to the obligatory pathogenic species of the Mycobacterium tuberculosis complex and Mycobacterium leprae, the genus Mycobacterium also includes conditionally pathogenic species that in rare cases can lead to the development of nontuberculous mycobacterial diseases. Because tuberculosis and mycobacteriosis

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service