Skip to Content
Merck
All Photos(1)

Key Documents

A66804

Sigma-Aldrich

3-Amino-2-naphthoic acid

technical grade, 80%

Synonym(s):

2-Amino-3-carboxynaphthalene, 2-Aminonaphthalene-3-carboxylic acid, 3-Amino-2-naphthalenecarboxylic acid, beta-Naphthylamine-3-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NC10H6CO2H
CAS Number:
Molecular Weight:
187.19
Beilstein:
744099
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

grade

technical grade

Quality Level

Assay

80%

form

powder, crystals or chunks

reaction suitability

reaction type: solution phase peptide synthesis

color

yellow to olive green

mp

212-215 °C (dec.) (lit.)

application(s)

peptide synthesis

SMILES string

Nc1cc2ccccc2cc1C(O)=O

InChI

1S/C11H9NO2/c12-10-6-8-4-2-1-3-7(8)5-9(10)11(13)14/h1-6H,12H2,(H,13,14)

InChI key

XFXOLBNQYFRSLQ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Understand sialic acid structure, function, signaling, and modifications. Easily find products for sialic acid research.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service