Skip to Content
Merck
All Photos(1)

Key Documents

935824

Sigma-Aldrich

Polyester-based biodegradable bioink

suitable for extrusion-based 3D bioprinting

Synonym(s):

SOLID INX

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200

Quality Level

form

solid

color

off-white

viscosity

10-30 Pa.s

mp

40-60 °C

gel strength

(Young′s modulus after UV crosslinking (Mpa) 150-200)

General description

The polyester-based biodegradable bioink is a photocrosslinkable, hydrophobic, and biocompatible bioink that combines the benefits of conventional stiff polyester materials with low temperature (<65°C) processability. This bioink has been used to generate and sustain 3D cellular structures of a variety of human tissues including cartilage (chondrocytes), blood vessels (HUVECs), smooh muscle cells (SMCs), and skin (HFFs).

Properties and Processing: The polyester-based biodegradable bioink is provided in a ready to print cartridge and is a semi-crystalline solid at room temperature with a melting emperature around 50°C and crystallization temperature of 25°C and displays Newtonian fluidic characteristics. The visocity of the bioink makes it suitable for an easy extrusion from the nozzlee during printing, resulting in a printable formulation in pneumatic 3D printers operating at pressures between 80-160 kPa. The ink is in molten state in the dispenser (~60°C), after which the printed constructs are solidified as a result of rapid cystrallization. The printed scaffold can then crosslinked via UV light.

Application

3D bioprinting
Tissue engineering
In vitro modeling

Features and Benefits

  • Easy printing - The bioink can be processed at low temperatures followed by photocrosslinking, making it ideal for hybrid scaffolds
  • Biocompatibility - Exceptional biocompatibility and no contact toxicity
  • Biodegradable - Fully biodegradable materials allows cellular remodeling of printed matrix
  • Easy handling - Delivered in a ready-to-use cartridge, ready to print after heating
  • UV curable - Efficient UV-based crosslinking
  • Reproducibility - Strict quality control
  • Mechanical integrity - Robust yet flexible polymer suitable for stiffer tissue engineering applications

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Multiphoton Lithography as a Promising Tool for Biomedical Applications
Greant, Coralie
Advanced Functional Materials , 33(39) (2023)
Olivier Guillaume et al.
Acta biomaterialia, 165, 72-85 (2022-03-16)
Since its inception, tissue engineering and regenerative medicine (TERM) has been relying on either scaffold-based or scaffold-free strategies. Recent reports outlined the possibility of a synergistic, convergence approach, referred to as the third TERM strategy, which could alleviate bottlenecks of

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service