Skip to Content
Merck
All Photos(1)

Key Documents

933015

Sigma-Aldrich

Wafer Tweezers

Style 43LB-4 Peek

Synonym(s):

SEM Sample Holder, Wafer Tweezers

Sign Into View Organizational & Contract Pricing

Select a Size

1 EACH
€94.90

€94.90


Estimated to ship on15 July 2025Details


Request a Bulk Order

Select a Size

Change View
1 EACH
€94.90

About This Item

UNSPSC Code:
41111761
NACRES:
NB.35

€94.90


Estimated to ship on15 July 2025Details


Request a Bulk Order

Looking for similar products? Visit Product Comparison Guide

General description

EMS techware

Application

Wafer Tweezers Style 43LB-8 Peek, made from Polyether ether ketone (PEEK), an Electrostatic discharge (ESD) safe soft, conductive fiber, with heat resistance up to 250 °C. Clean-room compatible. These tweezers are specially designed to handle different fragile and delicate materials of gallium or silicon wafers. They have a smooth polished non-glare surface with a satin finish. The tweezers have stopping pins turned up on lateral edges and graduated lower paddles prevent the wafer from slipping.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service