932949
Tweezer
High Precision, Style 1, Locking
Synonym(s):
Gripper, Tweezer
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
General description
EMS techware
Application
For the handling of delicate and fragile silicon wafers, scintillation discs, glass slides, coverslips. Made from antimagnetic/anti-acid steel with smooth surface. Self-closing.
Tip Width: 14.5 mm. Depth: 10.5 mm. Tip Lip: 9.0 mm.
Tip Width: 14.5 mm. Depth: 10.5 mm. Tip Lip: 9.0 mm.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service