Skip to Content
Merck
All Photos(1)

Key Documents

929816

Sigma-Aldrich

PCL-PEG-PCL diacrylate

new

average Mn (300-3400-300), poly(caprolactone)-b-poly(ethylene glycol)-b-poly(caprolactone) diacrylate

Synonym(s):

PCL-b-PEG-b-PCL, PEG hydrogel, acrylate-PCL-PEG-PCL-acrylate, biodegradable triblock copolymer

Sign Into View Organizational & Contract Pricing

Select a Size

929816-500MG
€440.00

€440.00


Please contact Customer Service for Availability


Select a Size

Change View
929816-500MG
€440.00

About This Item

Linear Formula:
C2H3O2(C6H10O2)x(C2H4O)y(OC6H10O)zOC2H3
UNSPSC Code:
12352200

€440.00


Please contact Customer Service for Availability

Quality Level

form

low-melting solid

mol wt

average Mn (300-3400-300)

contains

contains ≤ 300 ppm MEHQ as inhibitor

color

white to off-white

Ω-end

acrylate

α-end

acrylate

storage temp.

−20°C

SMILES string

O=C(OCCOC(CCCCCOC(C=C)=O)=O)CCCCCOC(C=C)=O

InChI

1S/C20H30O8/c1-3-17(21)25-13-9-5-7-11-19(23)27-15-16-28-20(24)12-8-6-10-14-26-18(22)4-2/h3-4H,1-2,5-16H2

InChI key

PCLZPCUCIICUMZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

PCL-PEG-PCL diacrylate, average Mn 300-3400-300, is a functionalized, amphiphilic triblock copolymer composed of a central hydrophilic, biocompatible poly(ethylene glycol) (PEG) block and two terminal hydrophobic, biodegradable poly(caprolactone) (PCL) blocks. PCL-PEG-PCL diacrylate can be crosslinked via free radical polymerization and/or photopolymerization to form a crosslinked hydrogel network. Hydrolysis of the esters of the PCL blocks permits degradation of the crosslinked networks.

Application

  • Tissue engineering
  • 3D printing
  • Drug delivery

Features and Benefits

  • PCL-PEG-PCL triblock copolymer
  • Hydrolytically degradable PCL blocks
  • Acrylate functional groups permit photopolymerization

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service