Skip to Content
Merck
All Photos(1)

Key Documents

926108

Sigma-Aldrich

(S)-RUCY-XylBINAP

Takasago

Synonym(s):

RuCl[(S)-daipena][(S)-xylbinap]

Sign Into View Organizational & Contract Pricing

Select a Size

250 MG
€72.00
1 G
€210.00
5 G
€575.00

€72.00


Please contact Customer Service for Availability


Select a Size

Change View
250 MG
€72.00
1 G
€210.00
5 G
€575.00

About This Item

Empirical Formula (Hill Notation):
C71H73ClN2O2P2Ru
CAS Number:
Molecular Weight:
1184.82
MDL number:
UNSPSC Code:
12352116
NACRES:
NA.22

€72.00


Please contact Customer Service for Availability

Quality Level

form

powder

reaction suitability

reagent type: ligand

functional group

phosphine

SMILES string

[Ru+]c9c(ccc(c9)OC)C(N)([C@@H](N)C(C)C)c%10ccc(cc%10)OC.P(c8cc(cc(c8)C)C)(c7cc(cc(c7)C)C)c1c(c6c(cc1)cccc6)c2c3c(ccc2P(c5cc(cc(c5)C)C)c4cc(cc(c4)C)C)cccc3.[Cl-]

InChI key

VBDGCLWXZBVBHR-UCPQXOCLSA-M

Related Categories

Application

(S)-RUCY-XylBINAP is a ruthenabicyclic complex that shows excellent activity in the asymmetric hydrogenation of ketones.

Legal Information

Product of Takasago.

For research and development purposes only. PCT/JP2010/072838
RUCY is a trademark of Takasago Intl. Corp.

related product

Product No.
Description
Pricing

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kazuhiko Matsumura et al.
Journal of the American Chemical Society, 133(28), 10696-10699 (2011-06-17)
A novel ruthenabicyclic complex with base shows excellent catalytic activity in the asymmetric hydrogenation of ketones. The turnover frequency of the hydrogenation of acetophenone reaches about 35,000 min(-1) in the best case, affording 1-phenylethanol in >99% ee. Several aliphatic and

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service