Skip to Content
Merck
All Photos(1)

Key Documents

911283

Sigma-Aldrich

Poly(ethylene glycol) linear dendron

carboxyl terminated, PEG Mn 10,000, generation 3

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C522H1000O273
Molecular Weight:
11645.36
UNSPSC Code:
51171641

form

solid

mol wt

PEG Mn 10,000

color

white to off-white

storage temp.

−20°C

General description

  • Polymer architecture: PEG-linear dendron
  • Generation: 3
  • PEG average Mn: 10,000
  • End Group Functionality: Carboxylic acid
  • No. Surface Groups: 8
  • Calculated Mol. Wt.: 11645.47 g/mol
  • Bis-MPA : 2,2-Bis(hydroxymethyl)propionic acid

Application

Bis-MPA dendritic molecules are based on 2,2-bismethylolpropionicacid (bis-MPA) building blocks. The hydrolyzable linker allows for degradation into byproducts that can be reabsorbed by the body or into excretable non-toxic small molecules. This dendritic polymer is biocompatible, biodegradable, and suitable for biomedical applications. PEG-core dendrons feature excellent solubility in water and a high degree of functionality per PEG chain, enable use for advanced drug delivery systems or for drug targeting. The carboxylic acid-functionalized corona can be readily used for bioconjugation reactions.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chemistry of multifunctional polymers based on bis-MPA and their cutting-edge applications.
Garcia-Gallego S, et al.
Progress in Polymer Science, 48, 85-110 (2015)
Chemistry of multifunctional polymers based on bis-MPA and their cutting-edge applications.
Garcia-Gallego S, et al
Progress in Polymer Science, 48, 85-110 (2015)
Anna Carlmark et al.
Chemical Society reviews, 42(13), 5858-5879 (2013-05-01)
Dendritic polymers are highly branched, globular architectures with multiple representations of functional groups. These nanoscale organic frameworks continue to fascinate researchers worldwide and are today under intensive investigation in application-driven research. A large number of potential application areas have been

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service