Skip to Content
Merck
All Photos(1)

Key Documents

901625

Sigma-Aldrich

Buffered oxide etchant (BOE) 10:1 with surfactant

Synonym(s):

BHF, Buffered HF

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161700
NACRES:
NA.23

form

liquid

General description

Buffered oxide etchant (BOE) is a wet etchant used in microfabrication. Its primary use is in etching thin films of silicon dioxide (SiO2) or silicon nitride (Si3N4). It is a mixture of a buffering agent, such as ammonium fluoride (NH4F), and hydrofluoric acid (HF). Concentrated HF etches silicon dioxide too quickly for good process control and also peels photoresist used in lithographic patterning.

Application

Buffered oxide etchant (BOE) 10:1 with surfactant can be potentially used in the etching of titanium carbide, which is used in the fabrication of microelectromechanical systems (MEMS).[1] It may be used in the etching of spin-on-dopant (SOD) for the development of conductor-insulator-conductor tunneling diodes.[2] It may also be used to enhance the surface of fused quartz devices.[3]

Pictograms

Skull and crossbonesCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Dermal - Acute Tox. 3 Inhalation - Acute Tox. 3 Oral - Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

6.1B - Non-combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Study on surface roughness improvement of Fused Quartz after thermal and chemical post-processing
2016 IEEE International Symposium on Inertial Sensors and Systems, 31(8), 101-104 (2016)
New process development for planar-type CIC tunneling diodes
Choi K, et al.
IEEE Electron Device Letters, 31(8), 809-811 (2010)
Integration of wear-resistant titanium carbide coatings into MEMS fabrication processes
Radhakrishnan G, et al.
Tribology Letters, 8(2-3), 133-137 (2000)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service