Skip to Content
Merck
All Photos(1)

Key Documents

900732

p-Xylene-(dimethyl-d6)

≥98 atom % D, ≥99% (CP)

Synonym(s):

1,4-Dimethyl-d6-benzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H4(CD3)2
CAS Number:
Molecular Weight:
112.20
MDL number:
UNSPSC Code:
12352116

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

≥98 atom % D

Assay

≥99% (CP)

form

liquid

mol wt

111.22

refractive index

n20/D 1.4944 (lit.)

bp

135.4 °C (lit.)

density

0.915 g/mL at 25 °C (lit.)

mass shift

M+6

SMILES string

[2H]C([2H])([2H])c1ccc(cc1)C([2H])([2H])[2H]

InChI

1S/C8H10/c1-7-3-5-8(2)6-4-7/h3-6H,1-2H3/i1D3,2D3

InChI key

URLKBWYHVLBVBO-WFGJKAKNSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Aquatic Chronic 3 - Asp. Tox. 1 - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

77.0 °F - closed cup

Flash Point(C)

25.00 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service