Skip to Content
Merck
All Photos(1)

Key Documents

900643

Sigma-Aldrich

Phosphazene base P2-Et

ZerO2®, ≥98.0%

Synonym(s):

Degassed and low oxygen phosphazene base, Phosphazene base ZerO2®, 1-Ethyl-2,2,4,4,4-pentakis(dimethylamino)-2λ5,4λ5-catenadi(phosphazene), Tetramethyl(tris(dimethylamino)phosphoranylidene)phosphorictriamid-Et-imin

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(CH3)2N]3P=NP(=NC2H5)[N(CH3)2]2
CAS Number:
Molecular Weight:
339.40
Beilstein:
7485991
MDL number:
UNSPSC Code:
12352000
grade:
ZerO2®

grade

ZerO2®

Assay

≥98.0%

form

liquid

refractive index

n20/D 1.492 (lit.)

bp

96 °C/0.05 mmHg (lit.)

density

1.02 g/mL at 25 °C (lit.)

functional group

amine

SMILES string

CCN=P(N=P(N(C)C)(N(C)C)N(C)C)(N(C)C)N(C)C

InChI

1S/C12H35N7P2/c1-12-13-20(15(2)3,16(4)5)14-21(17(6)7,18(8)9)19(10)11/h12H2,1-11H3

InChI key

CFUKEHPEQCSIOM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

ZerO2 products are rigorously degassed with highly pure inert gas providing solvents and solutions (anhydrous if specified) with very low residual oxygen content.

Legal Information

ZerO2 is a registered trademark of Sigma-Aldrich Co. LLC

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service