Skip to Content
Merck
All Photos(1)

Documents

806897

Sigma-Aldrich

ND3/He(RG) Gas Mixture

ratio (1:49), 99 atom % D

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352002

isotopic purity

99 atom % D

form

gas

composition

ratio (1:49)

Related Categories

Packaging

All gas volumes are quoted at Standard Temperature and Pressure (STP) and not at Normal Temperature and Pressure (NTP). One STP Liter is 7.7% more gas than one NTP Liter.
This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Aquatic Acute 1 - Eye Dam. 1 - Press. Gas Compr. Gas - Skin Corr. 1B

Storage Class Code

2A - Gases

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service