Skip to Content
Merck
All Photos(2)

Documents

804088

Sigma-Aldrich

SnAP 2-Spiro-(4-Pip) M Reagent

Synonym(s):

tert-butyl 4-(aminomethyl)-4-((tributylstannyl)methoxy)piperidine-1-carboxylate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C24H50N2O3Sn
Molecular Weight:
533.38
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

form

liquid

refractive index

n/D 1.492

density

1.146

storage temp.

−20°C

SMILES string

CCCC[Sn](CCCC)(COC1(CCN(C(OC(C)(C)C)=O)CC1)CN)CCCC

InChI

1S/C12H23N2O3.3C4H9.Sn/c1-11(2,3)17-10(15)14-7-5-12(9-13,16-4)6-8-14;3*1-3-4-2;/h4-9,13H2,1-3H3;3*1,3-4H2,2H3;

InChI key

LEBKPKYDLQUOJO-UHFFFAOYSA-N

General description

SnAP (Sn amino protocol) reagents have been developed by the Bode group SnAP reagents are air and moisture stable. They can be prepared by simple reactions with cheap raw materials. SnAP 2-Spiro-(4-Pip) M, also referred to as SnAP 2-spiro-(4-piperdine) morpholine, is a regioisomeric SnAP reagent. This reagent can be prepared from N-Boc piperidone.

Application

SnAP Reagents provide a one-step route, in tandem with various aldehyde substrates, to saturated N-heterocycles. The synthesis of N-heterocycles through SnAP Reagents requires mild reaction conditions, and aldehydes bearing aryl, heteroaryl, glyoxyl, aliphatic, and halogenated groups are well tolerated. This product was introduced in collaboration with the Bode Research Group
SnAP 2-Spiro-(4-Pip) M Reagent may be used for the preparation of unprotected, saturated N-heterocyclic compounds, C-substituted bicyclic and spirocyclic morpholines.

Automate your N-heterocycle formation with Synple Automated Synthesis Platform (SYNPLE-SC002)

related product

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Oral - Acute Tox. 4 Dermal - Aquatic Acute 1 - Aquatic Chronic 1 - Eye Irrit. 2 - Repr. 1B - Skin Irrit. 2 - STOT RE 1

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

>230.0 °F

Flash Point(C)

> 110 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kimberly Geoghegan et al.
Organic letters, 17(8), 1934-1937 (2015-03-31)
The precise placement of C-substituents on bicyclic and spirocyclic N-heterocycles is readily achieved by the combination of aldehydes and new SnAP reagents. The substituted SnAP reagents are readily prepared from simple starting materials and couple with a variety of aromatic
SnAP Reagents for a Cross-Coupling Approach to the One-Step Synthesis of Saturated N-Heterocycles.
Luescher MU, et al.
Aldrichimica Acta, 48(2), 43-48 (2015)
Cam-Van T Vo et al.
Nature chemistry, 6(4), 310-314 (2014-03-22)
Interest in saturated N-heterocycles as scaffolds for the synthesis of bioactive molecules is increasing. Reliable and predictable synthetic methods for the preparation of these compounds, especially medium-sized rings, are limited. We describe the development of SnAP (Sn amino protocol) reagents

Protocols

SnAP Reagents facilitate synthesis of saturated N-heterocycles for diverse structures.

SnAP Reagents facilitate synthesis of saturated N-heterocycles for diverse structures.

SnAP Reagents facilitate synthesis of saturated N-heterocycles for diverse structures.

SnAP Reagents facilitate synthesis of saturated N-heterocycles for diverse structures.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service