Skip to Content
Merck
All Photos(1)

Documents

803707

Sigma-Aldrich

Pd/graphene nanocomposite

10 mg/mL, dispersion in acetone

Synonym(s):

Pd decorated highly conductive graphene

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.23

description

Loading Density: 10%-50% (Pd nanocrystal) by TEM
Resistance: <103 Ω/sq (graphene)

form

dispersion

composition

acetone, ~80 wt. %
graphene, 6-10%
Pd nanoparticle, 2-6%

concentration

10 mg/mL

thickness

1-10 nm , graphene

particle size

5-50 nm (Pd nanocrystal)

storage temp.

2-8°C

Application

  • Perfect for transmission electron microscopy imaging standard.
  • A catalytic material to be used in fuel cells, CH4 gas sensor.

Legal Information

Product of Ocean Nanotech, LLC.

Pictograms

FlameExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 2 - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

41.0 °F

Flash Point(C)

5 °C


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Interface induced hydrogen sensing in Pd nanoparticle/graphene composite layers.
Kumar et al.
Sens. Actuators, 209, 919-926 (2015)
Pd/graphene nanocomposite as highly active catalyst for the Heck reactions Performance toward Methanol Electrooxidation.
Wang et al.
Applied Catalysis A: General, 489, 188-192 (2015)

Articles

Graphene is a unique two-dimensional (2D) structure of monolayer carbon atoms packed into a dense honeycomb crystal that has attracted great interest due to its diverse and fascinating properties.

Since its discovery little more than a decade ago,1 the two-dimensional (2D) allotrope of carbon—graphene—has been the subject of intense multidisciplinary research efforts.

Since its discovery little more than a decade ago,1 the two-dimensional (2D) allotrope of carbon—graphene—has been the subject of intense multidisciplinary research efforts.

Since its discovery little more than a decade ago,1 the two-dimensional (2D) allotrope of carbon—graphene—has been the subject of intense multidisciplinary research efforts.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service