Skip to Content
Merck
All Photos(1)

Key Documents

79785

Sigma-Aldrich

Phthalimide DBU salt

≥98.0%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C8H5NO2 · C9H16N2
CAS Number:
Molecular Weight:
299.37
Beilstein:
4343711
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥98.0%

mp

134-137 °C

SMILES string

O=C1NC(=O)c2ccccc12.C3CCN4CCCN=C4CC3

Other Notes

Reagent for Gabriel syntheses of amines soluble in CH2Cl2

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J. Wengel et al.
Synthesis, 829-829 (1989)
J Lau et al.
Acta chemica Scandinavica (Copenhagen, Denmark : 1989), 45(6), 616-620 (1991-07-01)
Mercuric-catalyzed hydrolysis of acetylated L-rhamnal 1 gives an alpha,beta-unsaturated aldehyde 2. 1,4-Addition of DBU-phthalimide salt with concomitant acetyl shift resulted in L-ribo and L-arabino isomers of 5-O-acetyl-2,3,6-trideoxy-3-phthalimido-hexofuranose 3 and 4. After acetylation at the anomeric center, coupling with silylated thymine

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service