790885
(R)-3,3′-Dibromo-1,1′-binaphthyl-2,2′-disulfonimide toluene complex
97%
Synonym(s):
(11bR)-2,6-Dibromodinaphtho[2,1-d:1′,2′-f][1,3,2]dithiazepine- 3,3,5,5-tetraoxide
About This Item
Recommended Products
Assay
97%
form
solid
reaction suitability
reaction type: Aldol Reaction
reaction type: Allylation
reaction type: Carbonyl/Imine Addition
reaction type: Friedel-Crafts Alkylation
mp
306-310 °C
SMILES string
CC1=CC=CC=C1.BrC2=CC(C=CC=C3)=C3C(C4=C(C=CC=C5)C5=CC(Br)=C4S6(=O)=O)=C2S(N6)(=O)=O.BrC7=CC(C=CC=C8)=C8C(C9=C(C=CC=C%10)C%10=CC(Br)=C9S%11(=O)=O)=C7S(N%11)(=O)=O
InChI
1S/2C20H11Br2NO4S2.C7H8/c2*21-15-9-11-5-1-3-7-13(11)17-18-14-8-4-2-6-12(14)10-16(22)20(18)29(26,27)23-28(24,25)19(15)17;1-7-5-3-2-4-6-7/h2*1-10,23H;2-6H,1H3
InChI key
WQYRPPIFPJBANM-UHFFFAOYSA-N
Application
A Powerful Chiral Counteranion Motif for Asymmetric Catalysis
Chiral Sulfonimide as a Brønsted Acid Organocatalyst for Asymmetric Friedel-Crafts Alkylation of Indoles with Imines
A href="http://onlinelibrary.wiley.com/doi/10.1002/ejoc.201300956/abstract" target="_blank">Chiral N-Fluorodibenzenesulfonimide Analogues for Enantioselective Electrophilic Fluorination and Oxidative Fluorination
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Asymmetric counteranion-directed catalysis (ACDC) has a number of valuable applications in enantioselective synthesis.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service