Skip to Content
Merck
All Photos(1)

Key Documents

778710

Sigma-Aldrich

2-(2-Furyl)-1,3-dioxolane

98%

Synonym(s):

2-(1,3-Dioxolan-2-yl)furan, 2-Furfural ethylene glycol acetal, Furfural ethylene acetal

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H8O3
CAS Number:
Molecular Weight:
140.14
Beilstein:
121450
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥97.5% (GC)
98%

storage temp.

−20°C

SMILES string

C1COC(O1)c2ccco2

InChI

1S/C7H8O3/c1-2-6(8-3-1)7-9-4-5-10-7/h1-3,7H,4-5H2

InChI key

FCACHSUJEMZCMK-UHFFFAOYSA-N

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service