Skip to Content
Merck
All Photos(1)

Key Documents

775339

Sigma-Aldrich

Dichloro[1,3-bis(2,6-diisopropylphenyl)-2-imidazolidinylidene][(2-isopropoxy)(5-pentafluorobenzoylamino)benzylidene]ruthenium(II)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C44H50Cl2F5N3O2Ru
CAS Number:
Molecular Weight:
919.85
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

form

powder

reaction suitability

core: ruthenium
reagent type: catalyst

mp

157-180 °C (decomposition)

storage temp.

2-8°C

SMILES string

Cl[Ru](=CC1=C(OC(C)C)C=CC(NC(C2=C(F)C(F)=C(F)C(F)=C2F)=O)=C1)(Cl)=C3N(C4=C(C(C)C)C=CC=C4C(C)C)CCN3C5=C(C(C)C)C=CC=C5C(C)C

InChI

1S/C27H38N2.C17H12F5NO2.2ClH.Ru/c1-18(2)22-11-9-12-23(19(3)4)26(22)28-15-16-29(17-28)27-24(20(5)6)13-10-14-25(27)21(7)8;1-7(2)25-10-5-4-9(6-8(10)3)23-17(24)11-12(18)14(20)16(22)15(21)13(11)19;;;/h9-14,18-21H,15-16H2,1-8H3;3-7H,1-2H3,(H,23,24);2*1H;/q;;;;+2/p-2

InChI key

XOFWGECEXOWWQY-UHFFFAOYSA-L

Legal Information

Umicore M7 catalysts are sold in cooperation with Umicore and OmegaCat Systems for research purposes only.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service