Skip to Content
Merck
All Photos(1)

Key Documents

772615

Sigma-Aldrich

2,2-Difluoro-1-propanol

95%

Synonym(s):

2,2-Difluoropropanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C3H6F2O
CAS Number:
Molecular Weight:
96.08
UNSPSC Code:
12352100
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.349

density

1.164 g/mL at 25 °C

InChI

1S/C3H6F2O/c1-3(4,5)2-6/h6H,2H2,1H3

InChI key

CKLONJANQGBREW-UHFFFAOYSA-N

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

88.0 °F - closed cup

Flash Point(C)

31.1 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service