Skip to Content
Merck
All Photos(1)

Key Documents

769487

Sigma-Aldrich

Zirconium(IV) oxide

pellets, diam. × thickness 13.5 mm × 9.5 mm

Synonym(s):

Zirconia

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ZrO2
CAS Number:
Molecular Weight:
123.22
EC Number:
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.23

form

pellets

diam. × thickness

13.5 mm × 9.5 mm

impurities

≤0.1% PBB (polybrominated biphenyls)
≤0.1% PBDE (polybrominated diphenyl ethers)

bp

5000 °C (lit.)

mp

2700 °C (lit.)

density

5.89 g/mL at 25 °C (lit.)

cation traces

Cd: ≤0.01%
Co: ≤0.0005%
Cr: ≤0.002%
Cu: ≤0.001%
Fe: ≤0.01%
Hg: ≤0.1%
Pb: ≤0.1%
Ti: ≤0.2%
V: ≤0.005%

SMILES string

O=[Zr]=O

InChI

1S/2O.Zr

InChI key

MCMNRKCIXSYSNV-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Zirconium Dioxide, ZrO2, is a high refractive index, low absorption material used for optical coatings in the near-UV to IR spectral regions.

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A high-performance spectrally-selective solar absorber based on a yttria-stabilized zirconia cermet with high-temperature stability
Cao, F., et al.
Energy & Environmental Science, 8(10), 3040-3048 (2015)
Silica?zirconia alkali-resistant coatings by sol?gel route
Dhere, SL.
Current Science, 108(9), 1647-1652 (2015)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service