Skip to Content
Merck
All Photos(1)

Documents

763497

Sigma-Aldrich

N,N,N′,N′-Tetrakis(4-methoxyphenyl)benzidine

98%

Synonym(s):

MeO-TPD

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C40H36N2O4
CAS Number:
Molecular Weight:
608.72
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

98%

form

powder

mp

190-195 °C

λmax

353 nm in chloroform

SMILES string

COc1ccc(cc1)N(c2ccc(OC)cc2)c3ccc(cc3)-c4ccc(cc4)N(c5ccc(OC)cc5)c6ccc(OC)cc6

InChI

1S/C40H36N2O4/c1-43-37-21-13-33(14-22-37)41(34-15-23-38(44-2)24-16-34)31-9-5-29(6-10-31)30-7-11-32(12-8-30)42(35-17-25-39(45-3)26-18-35)36-19-27-40(46-4)28-20-36/h5-28H,1-4H3

InChI key

WPUSEOSICYGUEW-UHFFFAOYSA-N

Application

This materials is a popular air stable hole transport material for OPV devices. It is often doped to improve mobility and performance. IT also increases the brightness of OLED devices (upto 14300 cd/m2 at 13V).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Investigation of C60F36 as low-volatility p-dopant in organic optoelectronic devices
Meerheim, R.;
Journal of Applied Physics, 109, 103102/1-103102/6 (2011)
Optical and Electrical Properties of OLED with the Structure of ITO/ m-MTDATA/Meo-TPD/Alq3/LiF/Al
Gui, T. L.; et al.
Journal of Physics. Conference Series, 276, 012076/1-012076/4 (2011)

Articles

Organic Light-emitting Diodes (OLEDs) are solid-state devices that transform electrical energy into light. OLEDs are considered the next generation technology for high-resolution flexible displays and solid state lighting, attracting intense scientific and industrial interest.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service