Skip to Content
Merck
All Photos(1)

Documents

761478

Sigma-Aldrich

N,N′-Bis(phenanthren-9-yl)-N,N′-bis(phenyl)-benzidine

99% (HPLC)

Synonym(s):

N,N′-Di-[(9-phenanthrenyl)-N,N′-diphenyl]-1,1′-biphenyl-4,4′-diamine, PAPB

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C52H36N2
CAS Number:
Molecular Weight:
688.86
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

99% (HPLC)

form

powder

mp

287-293 °C

absorption

115696 at 256 nm
30624 at 297 nm
42143 at 338 nm

SMILES string

c1ccc(cc1)N(c2ccc(cc2)-c3ccc(cc3)N(c4ccccc4)c5cc6ccccc6c7ccccc57)c8cc9ccccc9c%10ccccc8%10

InChI

1S/C52H36N2/c1-3-17-41(18-4-1)53(51-35-39-15-7-9-21-45(39)47-23-11-13-25-49(47)51)43-31-27-37(28-32-43)38-29-33-44(34-30-38)54(42-19-5-2-6-20-42)52-36-40-16-8-10-22-46(40)48-24-12-14-26-50(48)52/h1-36H

InChI key

LBFXFIPIIMAZPK-UHFFFAOYSA-N

Application

This material is a common hole transport material for OLED devices.

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Guangping Huang et al.
International journal of surgery (London, England), 61, 11-16 (2018-12-01)
The aim of the study was to introduce a novel percutaneous technique for the treatment of pelvic ring injuries using a percutaneous anterior pelvic bridge (PAPB) with K-wire. From December 2010 to November 2016, a prospective study of 86 patients
Mehmet Lütfi Yola et al.
Nanoscale, 12(38), 19824-19832 (2020-09-24)
Antigen galectin-3 (GL-3), a member of β-galactoside proteins indicates cardiac fibrosis and is a significant biomarker for monitoring heart failure risk and death risk. In this study, a novel sensitive amperometric method for antigen GL-3 detection is developed based on
Jia-Da Y.; et al.
Organic Electronics, 10, 1610-1610 (2009)
Robert B Denman et al.
Neuroscience letters, 572, 38-43 (2014-05-06)
Changes in cytoplasmic pH are known to regulate diverse cellular processes and influence neuronal activities. In neurons, the intracellular alkalization is shown to occur after stimulating several channels and receptors. For example, it has previously demonstrated in P19 neurons that
Manlong Xu et al.
Journal of pharmacological and toxicological methods, 71, 1-7 (2014-12-03)
PrestoBlue is a new resazurin based reagent to assess cell viability and cytotoxicity. It is claimed to be a fast and highly sensitive assay. Here, we compared PrestoBlue, alamarBlue, and 3-[4,5-dimethylthiazol-2-yl]-2,5-diphenyl tetrazonium bromide (MTT) in assessing cell viability of human

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service