Skip to Content
Merck
All Photos(1)

Key Documents

760633

Sigma-Aldrich

4′-Bromo-4-mercaptobiphenyl

Synonym(s):

4′-Bromo-[1,1′-biphenyl]-4-thiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H9BrS
CAS Number:
Molecular Weight:
265.17
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

solid

mp

149-157 °C

storage temp.

2-8°C

SMILES string

Sc1ccc(cc1)-c2ccc(Br)cc2

InChI

1S/C12H9BrS/c13-11-5-1-9(2-6-11)10-3-7-12(14)8-4-10/h1-8,14H

InChI key

JHPVDDGUGQROJG-UHFFFAOYSA-N

Application

It is used in functionalization of nanoparticles and assists in formimg self assembled monolayers (SAM)

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

212.0 °F

Flash Point(C)

100 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Novel One-Phase Synthesis of Thiol-Functionalized Gold,
Palladium, and Iridium Nanoparticles Using Superhydride
Yee CK, et al
Langmuir, 15, 3486-3491 (1999)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service