Skip to Content
Merck
All Photos(1)

Documents

753882

Sigma-Aldrich

CdSeS/ZnS alloyed quantum dots

COOH functionalized, fluorescence λem 630 nm, 6 nm diameter, 1 mg/mL in H2O

Synonym(s):

QD carboxyl functionalized, Fluorescent Nanocrystals, Trilite

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CdSxSe1-x/ZnS
UNSPSC Code:
26111700
NACRES:
NA.23

form

liquid

concentration

1 mg/mL in H2O

refractive index

n20/D 1.496 (lit.)

diameter

6 nm

bp

110-111 °C (lit.)

fluorescence

λem 630 nm

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Application

These Photo and Thermostable Quantum Dot materials have comparable fluorescence and molecular kinetics across all colors. Narrow and sharp emission peaks result in less overlap of peaks when multiplexing.

Analysis Note

Full Width at Half Maximum: 30 - 40nm

Legal Information

For use in optical electronics research only.
Product of CytoDiagnostics
Trilite is a trademark of Cytodiagnostics Inc.

Pictograms

Environment

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 2

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Dye-sensitized solar cells as a promising, low-cost photovoltaic technology.

Dye-sensitized solar cells as a promising, low-cost photovoltaic technology.

Dye-sensitized solar cells as a promising, low-cost photovoltaic technology.

Dye-sensitized solar cells as a promising, low-cost photovoltaic technology.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service