Skip to Content
Merck
All Photos(1)

Documents

748870

Sigma-Aldrich

9-Mercaptofluorene

Synonym(s):

9-Fluorenethiol, 9-Fluorenthiol, 9-Mercapto-fluoren, Fluorene-9-thiol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C13H10S
CAS Number:
Molecular Weight:
198.28
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

form

solid

mp

103-107 °C

SMILES string

SC1c2ccccc2-c3ccccc13

InChI

1S/C13H10S/c14-13-11-7-3-1-5-9(11)10-6-2-4-8-12(10)13/h1-8,13-14H

InChI key

GNURAXCIBMZJOG-UHFFFAOYSA-N

Application

This novel material is capable of self-assembly onto gold surfaces. The fluorene group is a precursor for the synthesis of luminophore materials that have longer lifespans and more efficient luminescence.

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cheol, H. C.; Joo, S. H.; Kim, K.; Jin, J. I.; Shin, H. W.; Kim, Y. R.
Macromolecules, 38, 6336-6336 (2005)

Articles

Recent research highlights tunable properties of inorganic nanoparticles, driving interest in optoelectronics.

Recent research highlights tunable properties of inorganic nanoparticles, driving interest in optoelectronics.

Recent research highlights tunable properties of inorganic nanoparticles, driving interest in optoelectronics.

Recent research highlights tunable properties of inorganic nanoparticles, driving interest in optoelectronics.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service