Skip to Content
Merck
All Photos(1)

Documents

74885

Sigma-Aldrich

Sodium 1-octanesulfonate monohydrate

≥97.0% (T)

Synonym(s):

1-Octanesulfonic acid sodium salt monohydrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)6CH2SO3Na · H2O
CAS Number:
Molecular Weight:
234.29
Beilstein:
3574241
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

≥97.0% (T)

form

sheet

mp

≥300 °C (lit.)

solubility

H2O: 0.1 g/mL, clear, colorless

SMILES string

O.[Na+].CCCCCCCCS([O-])(=O)=O

InChI

1S/C8H18O3S.Na.H2O/c1-2-3-4-5-6-7-8-12(9,10)11;;/h2-8H2,1H3,(H,9,10,11);;1H2/q;+1;/p-1

InChI key

MBURIAHQXJQKRE-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

Sodium 1-octanesulfonate is an ion-pairing agent mainly used in chromatographic mobile phase.

Application


  • Effect of Pretreatment Processes on Biogenic Amines Content and Some Bioactive Compounds in Hericium erinaceus Extract: This study details the mobile phase composition for chromatographic analysis, including Sodium 1-octanesulfonate monohydrate (N Makhamrueang et al., 2021).

  • What the New USP Chapter 621 Guidelines Mean for Your HPLC Analysis: Describes the use of Sodium 1-octanesulfonate in HPLC mobile phase preparation to meet new pharmacopeia standards (K Gleerup, 2023).

  • Simultaneous determination of fangchinoline and tetrandrine in Qi-Fang-Xi-Bi-granules by RP-HPLC: Utilizes Sodium 1-octanesulfonate as part of the mobile phase in the RP-HPLC method developed for analyzing traditional Chinese medicine (XG Lu et al., 2015).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Strong induction of phytochelatin synthesis by zinc in marine green alga, Dunaliella tertiolecta.
Hirata K, et al.
Journal of Bioscience and Bioengineering, 92(1), 24-29 (2001)
N M Kovalchuk et al.
Journal of colloid and interface science, 459, 250-256 (2015-08-25)
Mixed solutions of cationic and anionic surfactants show considerable synergism in their wetting behaviour, but their spreading is affected considerably by the phase separation processes. The valuable information about wetting properties of synergetic mixtures can be obtained by using mixtures
Maria Addolorata Saracino et al.
Journal of pharmaceutical and biomedical analysis, 104, 122-129 (2014-12-17)
A new microextraction by packed sorbent (MEPS) procedure coupled to a high-performance liquid chromatographic method has been developed for quantitation of catecholamines (i.e. norepinephrine, epinephrine and dopamine) in innovative biological samples, namely dried plasma and urine spots. Analyses were carried

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service