Skip to Content
Merck
All Photos(1)

Key Documents

748668

Sigma-Aldrich

2,3-Dichloropyridine-4-boronic acid MIDA ester

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H9BCl2N2O4
Molecular Weight:
302.91
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:

Assay

95%

form

solid

mp

292-294 °C

SMILES string

CN1CC(=O)OB(OC(=O)C1)c2ccnc(Cl)c2Cl

InChI

1S/C10H9BCl2N2O4/c1-15-4-7(16)18-11(19-8(17)5-15)6-2-3-14-10(13)9(6)12/h2-3H,4-5H2,1H3

InChI key

AKCTWCKWACJNIV-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service