Skip to Content
Merck
All Photos(1)

Key Documents

736902

Sigma-Aldrich

1-(Boc-amino)adamantane

95%

Synonym(s):

N-t-Butoxycarbonyl-1-adamantylamine, N-Boc-1-adamantylamine, tert-Butyl N-tricyclo[3.3.1.13,7]dec-1-ylcarbamate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H25NO2
CAS Number:
Molecular Weight:
251.36
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
powder
Assay:
95%

Assay

95%

form

powder

color

white

mp

110-115 °C

storage temp.

−20°C

SMILES string

CC(C)(C)OC(=O)NC12C[C@H]3C[C@H](C[C@H](C3)C1)C2

InChI

1S/C15H25NO2/c1-14(2,3)18-13(17)16-15-7-10-4-11(8-15)6-12(5-10)9-15/h10-12H,4-9H2,1-3H3,(H,16,17)/t10-,11+,12-,15-

InChI key

LSVGBVIPHLXQPG-WUQLGEGHSA-N

Pictograms

Environment

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Acute 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service