Skip to Content
Merck
All Photos(2)

Key Documents

729582

Sigma-Aldrich

9-Benzyl-9H-carbazole-3-boronic acid pinacol ester

95%

Synonym(s):

9-benzyl-3-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-9H-carbazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H26BNO2
Molecular Weight:
383.29
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

solid

mp

146-151 °C

SMILES string

CC1(C)OB(OC1(C)C)c2ccc3n(Cc4ccccc4)c5ccccc5c3c2

InChI

1S/C25H26BNO2/c1-24(2)25(3,4)29-26(28-24)19-14-15-23-21(16-19)20-12-8-9-13-22(20)27(23)17-18-10-6-5-7-11-18/h5-16H,17H2,1-4H3

InChI key

UMWJZMUSYJMTOI-UHFFFAOYSA-N

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service