Skip to Content
Merck
All Photos(1)

Key Documents

727105

Sigma-Aldrich

(R)-(−)-2-Aminoheptane

ChiPros®, produced by BASF, 99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3(CH2)4CH(NH2)CH3
CAS Number:
Molecular Weight:
115.22
Beilstein:
4652352
MDL number:
UNSPSC Code:
12352116
PubChem Substance ID:

Assay

≥98.5% (GC)
99%

form

liquid

optical purity

enantiomeric excess: ≥98.5%

refractive index

n20/D 1.419 (lit.)

bp

142-144 °C (lit.)

density

0.766 g/mL at 25 °C (lit.)

SMILES string

CCCCC[C@@H](C)N

InChI

1S/C7H17N/c1-3-4-5-6-7(2)8/h7H,3-6,8H2,1-2H3/t7-/m1/s1

InChI key

VSRBKQFNFZQRBM-SSDOTTSWSA-N

Looking for similar products? Visit Product Comparison Guide

Legal Information

ChiPros is a registered trademark of BASF SE

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Flam. Liq. 3 - Skin Corr. 1B - Skin Sens. 1 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

111.2 °F

Flash Point(C)

44 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service