Skip to Content
Merck
All Photos(1)

Key Documents

726230

Sigma-Aldrich

Boc-Orn-OH

≥98% (TLC)

Synonym(s):

Nα-tert-Butoxycarbonyl-L-ornithine, Nα-Boc-L-ornithine, N2-Boc-L-ornithine

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H20N2O4
CAS Number:
Molecular Weight:
232.28
Beilstein:
4313353
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98% (TLC)

form

powder

optical activity

[α]/D +24.0±2.0°, c = 1 in methanol

composition

carbon, 51.0-52.4%
nitrogen, 11.8-12.3%

reaction suitability

reaction type: Boc solid-phase peptide synthesis

application(s)

peptide synthesis

SMILES string

NCCC[C@H](NC(OC(C)(C)C)=O)C(O)=O

InChI

1S/C10H20N2O4/c1-10(2,3)16-9(15)12-7(8(13)14)5-4-6-11/h7H,4-6,11H2,1-3H3,(H,12,15)(H,13,14)/t7-/m0/s1

InChI key

AMPVNPYPOOQUJF-ZETCQYMHSA-N

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Skin Sens. 1

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service