72180
cis-Nerolidol
≥96.0% (GC)
Synonym(s):
cis-3,7,11-Trimethyl-1,6,10-dodecatrien-3-ol
About This Item
Recommended Products
Quality Level
Assay
≥96.0% (GC)
form
liquid
refractive index
n20/D 1.478 (lit.)
bp
70 °C/0.1 mmHg (lit.)
density
0.876 g/mL at 20 °C (lit.)
SMILES string
C\C(C)=C\CC\C(C)=C/CCC(C)(O)C=C
InChI
1S/C15H26O/c1-6-15(5,16)12-8-11-14(4)10-7-9-13(2)3/h6,9,11,16H,1,7-8,10,12H2,2-5H3/b14-11-
InChI key
FQTLCLSUCSAZDY-KAMYIIQDSA-N
Storage Class Code
10 - Combustible liquids
WGK
WGK 3
Flash Point(F)
204.8 °F - closed cup
Flash Point(C)
96 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service