Skip to Content
Merck
All Photos(1)

Key Documents

718610

Sigma-Aldrich

3-[(2-Boc-amino)acetamido]phenylboronic acid pinacol ester

97%

Synonym(s):

([3-(4,4,5,5-Tetramethyl-[1,3,2]dioxaborolan-2-yl)-phenylcarbamoyl]-methyl)carbamic acid tert-butyl ester, 3-[2-(tert-Butoxycarbonylamino)acetamido]phenylboronic acid pinacol ester, tert-Butyl([3-(4,4,5,5-tetramethyl-[1,3,2]dioxaborolan-2-yl)-phenylcarbamoyl]methylcarbamate)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C19H29BN2O5
Molecular Weight:
376.25
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

155-160 °C

functional group

amide
amine

SMILES string

CC(C)(C)OC(=O)NCC(=O)Nc1cccc(c1)B2OC(C)(C)C(C)(C)O2

InChI

1S/C19H29BN2O5/c1-17(2,3)25-16(24)21-12-15(23)22-14-10-8-9-13(11-14)20-26-18(4,5)19(6,7)27-20/h8-11H,12H2,1-7H3,(H,21,24)(H,22,23)

InChI key

IYTPRTWBNLJZLK-UHFFFAOYSA-N

Application

3-[(2-Boc-amino)acetamido]phenylboronic acid pinacol ester can be used to prepare a boronic acid-derived artificial receptor, which selectively binds to a neurotransmitter 3,4-dihydroxyphenylalanine (L-DOPA) in aqueous media and also inhibits the enzyme L-DOPA decarboxylase.[1]

Storage Class Code

11 - Combustible Solids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Design, synthesis and evaluation of a boronic acid based artificial receptor for l-DOPA in aqueous media
Ueno H, et al.
Chemical Communications (Cambridge, England), 49(88), 10403-10405 (2013)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service