Skip to Content
Merck
All Photos(1)

Key Documents

703486

Sigma-Aldrich

sec-Butylmagnesium chloride lithium chloride complex solution

1.2 M in THF

Synonym(s):

2-Butylmagnesium chloride lithium chloride complex, Turbo Grignard

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C4H9MgCl · LiCl
Molecular Weight:
159.27
UNSPSC Code:
12352001
PubChem Substance ID:
NACRES:
NA.22

form

liquid

reaction suitability

reaction type: Grignard Reaction

concentration

1.2 M in THF
14 % (w/w)

density

0.973 g/mL at 25 °C

SMILES string

[Li]Cl.CCC(C)[Mg]Cl

InChI

1S/C4H9.2ClH.Li.Mg/c1-3-4-2;;;;/h3H,4H2,1-2H3;2*1H;;/q;;;2*+1/p-2

InChI key

ZMDHAYOBNXOYEU-UHFFFAOYSA-L

Application

Reagent for Selective Metalations

Used as:
  • Functionalization of cyclopentene derivatives

Legal Information

Product of Albemarle US Inc

Signal Word

Danger

Hazard Classifications

Acute Tox. 4 Oral - Carc. 2 - Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - STOT SE 3 - Water-react 1

Target Organs

Central nervous system, Respiratory system

Supplementary Hazards

Storage Class Code

4.3 - Hazardous materials which set free flammable gases upon contact with water

WGK

WGK 1

Flash Point(F)

-6.7 °F

Flash Point(C)

-21.5 °C

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Articles

Salt additives increase both the rate and the efficiency of the Mg-halogen exchange reaction. The most effective reagents are generated with R-MgCl (R = i-Pr, s-Butyl) and 1.0 equiv of LiCl.

Salt additives increase both the rate and the efficiency of the Mg-halogen exchange reaction. The most effective reagents are generated with R-MgCl (R = i-Pr, s-Butyl) and 1.0 equiv of LiCl.

Salt additives increase both the rate and the efficiency of the Mg-halogen exchange reaction. The most effective reagents are generated with R-MgCl (R = i-Pr, s-Butyl) and 1.0 equiv of LiCl.

Salt additives increase both the rate and the efficiency of the Mg-halogen exchange reaction. The most effective reagents are generated with R-MgCl (R = i-Pr, s-Butyl) and 1.0 equiv of LiCl.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service