702846
Trimethylamine
anhydrous, cylinder, with 316SS needle valve, 99%
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(CH3)3N
CAS Number:
Molecular Weight:
59.11
Beilstein:
956566
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:
NACRES:
NA.22
grade
anhydrous
vapor density
2.09 (vs air)
vapor pressure
13.3 psi ( 21 °C)
Assay
99%
form
cylinder
autoignition temp.
374 °F
expl. lim.
11.6 %
bp
3-4 °C (lit.)
mp
−117 °C (lit.)
density
0.63 g/mL at 20 °C (lit.)
storage temp.
2-8°C
SMILES string
CN(C)C
InChI
1S/C3H9N/c1-4(2)3/h1-3H3
InChI key
GETQZCLCWQTVFV-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Packaging
Supplied in a Sure/Pac™ cylinder and has a 316 stainless steel needle valve with a male 1/4" NPTF outlet thread installed . Before using the cylinder, ensure that the valve is closed, then remove the galvanized steel hex cap that seals the outlet valve.
Compatible with the following:
Compatible with the following:
Legal Information
Aldrich is a registered trademark of Sigma-Aldrich Co. LLC
Sure/Pac is a trademark of Sigma-Aldrich Co. LLC
also commonly purchased with this product
Product No.
Description
Pricing
hose barb
Product No.
Description
Pricing
recommended
Product No.
Description
Pricing
regulator
Product No.
Description
Pricing
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Inhalation - Eye Dam. 1 - Flam. Gas 1A - Press. Gas Liquefied gas - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
2A - Gases
WGK
WGK 1
Flash Point(F)
19.4 °F - closed cup
Flash Point(C)
-7 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
M Al-Waiz et al.
Clinical pharmacology and therapeutics, 42(5), 588-594 (1987-11-01)
Trimethylamine (TMA) and its N-oxide (TMAO) are normal components of human urine. They are present in the diet and also derived from the enterobacterial metabolism of precursors such as choline. Dietary TMA is almost entirely metabolized to and excreted as
S L Hillier
American journal of obstetrics and gynecology, 169(2 Pt 2), 455-459 (1993-08-01)
Bacterial vaginosis is characterized microbiologically by replacement of the Lactobacillus-predominant vaginal flora by Gardnerella vaginalis, Bacteroides species, Mobiluncus species, and genital mycoplasmas. A standardized, laboratory-based diagnostic test for bacterial vaginosis is desirable in those instances in which a microscope is
Hyeryeong Jeon et al.
Macromolecular rapid communications, 33(11), 972-976 (2012-04-12)
Polydiacetylenes (PDAs), a family of conjugated polymers, are very intriguing materials in several aspects. Especially, the stimulus-induced apparent blue-to-red transition of the PDAs has led to the development of a variety of PDA-based chemosensors. In the current work, we synthesized
Flurin Cathomas et al.
Brain, behavior, and immunity, 50, 125-140 (2015-07-15)
The similarity between sickness behavior syndrome (SBS) in infection and autoimmune disorders and certain symptoms in major depressive disorder (MDD), and the high co-morbidity of autoimmune disorders and MDD, constitutes some of the major evidence for the immune-inflammation hypothesis of
Qian Li et al.
Current biology : CB, 23(1), 11-20 (2012-11-28)
Rodents use olfactory cues for species-specific behaviors. For example, mice emit odors to attract mates of the same species, but not competitors of closely related species. This implies rapid evolution of olfactory signaling, although odors and chemosensory receptors involved are
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service