Recommended Products
Assay
≥96%
form
powder
reaction suitability
reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: ligand
mp
67-71 °C
functional group
phosphine
SMILES string
[Fe].ClP(Cl)[C]1[CH][CH][CH][CH]1.ClP(Cl)[C]2[CH][CH][CH][CH]2
InChI
1S/2C5H4Cl2P.Fe/c2*6-8(7)5-3-1-2-4-5;/h2*1-4H;
InChI key
CSJDQGIPHURAPH-UHFFFAOYSA-N
Application
- Ferrocene-based bidentate phosphonite ligands for rhodium(I)-catalyzed enantioselective hydroformylation
- Metallocene-bridged bisphosphaalkene
Catalyst for synthesis of oligonuclear heterometallic complexes
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8B - Non-combustible corrosive hazardous materials
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service