Skip to Content
Merck
All Photos(1)

Key Documents

691011

Sigma-Aldrich

(S)-(−)-α-Allylalanine

≥98.0% (HPLC)

Synonym(s):

(S)-2-Amino-2-methyl-4-pentenoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H11NO2
CAS Number:
Molecular Weight:
129.16
MDL number:
UNSPSC Code:
12352200
eCl@ss:
32160406
PubChem Substance ID:
NACRES:
NA.22

Assay

≥98.0% (HPLC)

form

powder

optical purity

ee: ≥98.0% (HPLC)

reaction suitability

reaction type: solution phase peptide synthesis

impurities

≤15% water

application(s)

peptide synthesis

storage temp.

2-8°C

SMILES string

C[C@](N)(CC=C)C(O)=O

InChI

1S/C6H11NO2/c1-3-4-6(2,7)5(8)9/h3H,1,4,7H2,2H3,(H,8,9)/t6-/m0/s1

InChI key

QMBTZYHBJFPEJB-LURJTMIESA-N

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Thanh K Pham et al.
Bioorganic chemistry, 101, 104024-104024 (2020-07-07)
The N-capping box is a distinct helix-stabilizing motif frequently found in proteins. In this study, we examined a ruthenium-mediated intramolecular backbone to side chain macrocyclization as a rigidified mimicry of the N-capping box. Experimental data indicate that the 15-membered macrocycle

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service