Skip to Content
Merck
All Photos(1)

Documents

679291

Sigma-Aldrich

Triethoxyphenylsilane

deposition grade, ≥98%

Synonym(s):

Phenyltriethoxysilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5Si(OC2H5)3
CAS Number:
Molecular Weight:
240.37
Beilstein:
2940602
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

grade

deposition grade

vapor density

>1 (vs air)

Assay

≥98%

form

liquid

refractive index

n20/D 1.461 (lit.)

bp

112-113 °C/10 mmHg (lit.)

density

0.996 g/mL at 25 °C (lit.)

SMILES string

CCO[Si](OCC)(OCC)c1ccccc1

InChI

1S/C12H20O3Si/c1-4-13-16(14-5-2,15-6-3)12-10-8-7-9-11-12/h7-11H,4-6H2,1-3H3

InChI key

JCVQKRGIASEUKR-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameHealth hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Flam. Liq. 3 - STOT RE 2 Oral

Target Organs

Urinary bladder

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

128.3 °F - closed cup

Flash Point(C)

53.5 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Regulatory Listings

Regulatory Listings are mainly provided for chemical products. Only limited information can be provided here for non-chemical products. No entry means none of the components are listed. It is the user’s obligation to ensure the safe and legal use of the product.

EU REACH Annex XVII (Restriction List)

CAS No.

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yang Lv et al.
ACS applied materials & interfaces, 7(21), 11280-11285 (2015-05-15)
Percutaneous vertebroplasty (VP) and balloon kyphoplasty (BKP) are now widely used to treat patients who suffer painful vertebral compression fractures. In each of these treatments, a bone cement paste is injected into the fractured vertebral body/bodies, and the cement of
Yueer Yan et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 21(34), 12161-12170 (2015-07-16)
A clear and deep understanding of zeolite crystallization with the addition of organosilane is desirable for the reasonable design and preparation of hierarchical zeolites. Herein, the effects of different organosilanes on zeolite crystallization were systematically studied. It was found that
Frank M Hauser et al.
Forensic science international, 253, 55-63 (2015-06-06)
This paper deals with the synthesis and application of lanthanide complex doped nanobeads used as a luminescent fingerprint powder. Due to their special optical properties, namely a long emission lifetime, sharp emission profiles and large Stokes shifts, luminescent lanthanide complexes

Articles

Deposition Grade Silanes, fully characterized by chemical analysis and nuclear magnetic resonance (NMR) with greater than 98% purity, for Sol-Gel Processes.

Mesoporous materials, such as aerogels, offer advantages for practical hydrogen storage. They have large surface areas, open porosity, small pore sizes, and the ability to coat the surface with one or more compounds.

Mesoporous materials, such as aerogels, offer advantages for practical hydrogen storage. They have large surface areas, open porosity, small pore sizes, and the ability to coat the surface with one or more compounds.

Mesoporous materials, such as aerogels, offer advantages for practical hydrogen storage. They have large surface areas, open porosity, small pore sizes, and the ability to coat the surface with one or more compounds.

See All

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service