Skip to Content
Merck
All Photos(1)

Key Documents

666297

Sigma-Aldrich

(R)-(−)-3-Aminopiperidine dihydrochloride

97%

Synonym(s):

(R)-3-Piperidinamine dihydrochloride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H12N2 · 2HCl
CAS Number:
Molecular Weight:
173.08
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

190-195 °C
205-209 °C

SMILES string

Cl.Cl.N[C@@H]1CCCNC1

InChI

1S/C5H12N2.2ClH/c6-5-2-1-3-7-4-5;;/h5,7H,1-4,6H2;2*1H/t5-;;/m1../s1

InChI key

GGPNYXIOFZLNKW-ZJIMSODOSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service