Skip to Content
Merck
All Photos(1)

Key Documents

665746

Sigma-Aldrich

4-Pentylphenyl 4-pentylbenzoate

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H30O2
CAS Number:
Molecular Weight:
338.48
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.23

Assay

97%

form

liquid crystal

mp

34-38 °C

storage temp.

2-8°C

SMILES string

CCCCCc1ccc(OC(=O)c2ccc(CCCCC)cc2)cc1

InChI

1S/C23H30O2/c1-3-5-7-9-19-11-15-21(16-12-19)23(24)25-22-17-13-20(14-18-22)10-8-6-4-2/h11-18H,3-10H2,1-2H3

InChI key

VWDNHTVWLXZZEK-UHFFFAOYSA-N

Hazard Statements

Hazard Classifications

Aquatic Chronic 4

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service