Skip to Content
Merck
All Photos(1)

Documents

656437

Sigma-Aldrich

Methyl 2,6-dichloropyridine-3-carboxylate

97%

Synonym(s):

Methyl 2,6-dichloronicotinate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C7H5Cl2NO2
CAS Number:
Molecular Weight:
206.03
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

solid

mp

56-60 °C (lit.)

SMILES string

COC(=O)c1ccc(Cl)nc1Cl

InChI

1S/C7H5Cl2NO2/c1-12-7(11)4-2-3-5(8)10-6(4)9/h2-3H,1H3

InChI key

IFVVGOJYWCHRCT-UHFFFAOYSA-N

Application

Replacement of chlorides with fluorides using anhydrous nucleophilic fluoride salts generated from potassium fluoride and an arene transfer agent.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Haoran Sun et al.
Chemical communications (Cambridge, England), (5), 528-529 (2007-01-26)
Fluoride relay is used to generate exceptionally nucleophilic fluoride reagents from KF on a time scale commensurate with radiotracer synthesis.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service