Skip to Content
Merck
All Photos(1)

Documents

593834

Sigma-Aldrich

5-Aminolevulinic acid-3-13C hydrochloride

≥99 atom % 13C, ≥99% (CP)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2NCH2CO13CH2CH2CO2H · HCl
CAS Number:
Molecular Weight:
168.58
MDL number:
UNSPSC Code:
12352115
PubChem Substance ID:

isotopic purity

≥99 atom % 13C

Assay

≥99% (CP)

form

solid

mp

156 °C (dec.) (lit.)

mass shift

M+1

storage temp.

-10 to -25°C

SMILES string

Cl.NCC(=O)[13CH2]CC(O)=O

InChI

1S/C5H9NO3.ClH/c6-3-4(7)1-2-5(8)9;/h1-3,6H2,(H,8,9);1H/i1+1;

InChI key

ZLHFONARZHCSET-YTBWXGASSA-N

Related Categories

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service