Skip to Content
Merck
All Photos(1)

Key Documents

591793

Sigma-Aldrich

2-Methyl-2-nitropropane-15N

98 atom % 15N

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3C15NO2
Molecular Weight:
104.11
MDL number:
UNSPSC Code:
12352102
PubChem Substance ID:

isotopic purity

98 atom % 15N

bp

126-127 °C (lit.)

density

0.959 g/mL at 25 °C

mass shift

M+1

SMILES string

CC(C)(C)[15N+]([O-])=O

InChI

1S/C4H9NO2/c1-4(2,3)5(6)7/h1-3H3/i5+1

InChI key

AIMREYQYBFBEGQ-HOSYLAQJSA-N

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Flame

Signal Word

Danger

Hazard Statements

Hazard Classifications

Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

66.2 °F - closed cup

Flash Point(C)

19 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service