Skip to Content
Merck
All Photos(1)

Key Documents

58920

Sigma-Aldrich

L-Isoleucine methyl ester hydrochloride

≥98.0% (AT)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH(CH3)CH(NH2)COOCH3 · HCl
CAS Number:
Molecular Weight:
181.66
Beilstein:
3595132
EC Number:
MDL number:
UNSPSC Code:
12352200
eCl@ss:
32160406
PubChem Substance ID:

Assay

≥98.0% (AT)

optical activity

[α]20/D +27.0±1°, c = 2% in H2O

reaction suitability

reaction type: solution phase peptide synthesis

mp

96-100 °C

application(s)

peptide synthesis

SMILES string

Cl.CC[C@H](C)[C@H](N)C(=O)OC

InChI

1S/C7H15NO2.ClH/c1-4-5(2)6(8)7(9)10-3;/h5-6H,4,8H2,1-3H3;1H/t5-,6-;/m0./s1

InChI key

GGTBEWGOPAFTTH-GEMLJDPKSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

I B Rozenval'd
Tsitologiia, 32(3), 220-226 (1990-01-01)
Lysosomal inhibitors (amino acid methyl esters) and the platelet-derived growth factor stimulate the resting NIH 3T3 cells to enter the S period. Incubation of cells in the medium containing lysosomal inhibitors enhances protein accumulation and seemingly does not disrupt lysosomes.
Walter S Leal et al.
Journal of chemical ecology, 29(1), 15-25 (2003-03-22)
Three amino acid-derived compounds were identified in extracts from the pheromone glands of the scarab beetle Phyllophaga elenans, i.e., L-isoleucine methyl ester (LIME), N-formyl L-isoleucine methyl ester (For-LIME), and N-acetyl L-isoleucine methyl ester (Ac-LIME). The compounds were characterized from their
S C Alfieri et al.
Parasitology, 95 ( Pt 1), 31-41 (1987-08-01)
L-Amino acid esters, such as leucine methyl ester (Leu-OMe) can destroy intracellular as well as isolated amastigotes of Leishmania mexicana amazonensis by a mechanism which may involve ester hydrolysis by parasite enzymes. We show here that several other esters prevented
Paul S Robbins et al.
Journal of chemical ecology, 34(2), 205-214 (2008-01-24)
This study offers experimental evidence for the existence of three pheromone races of the northern genitalic form of Phyllophaga anxia: one race in which females produce and males respond mainly to L-valine methyl ester, a second producing and responding to
Yuta Kawasaki et al.
The Journal of experimental biology, 220(Pt 21), 4024-4034 (2017-09-08)
Two-day rhythms, referred to as circa'bi'dian rhythms, have been reported in humans and mosquitos. However, these rhythms only appear under constant conditions, and the functional mechanisms of 2-day rhythms were unknown. Here, we report clear circabidian rhythms of large black

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service