Skip to Content
Merck
All Photos(1)

Key Documents

588601

Sigma-Aldrich

Carbon dioxide-18O2

50 atom % 18O

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C18O2
CAS Number:
Molecular Weight:
48.01
EC Number:
MDL number:
UNSPSC Code:
12142200
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

50 atom % 18O

mp

-78 °C (subl.) (lit.)

mass shift

M+4

SMILES string

[18O]=C=[18O]

InChI

1S/CO2/c2-1-3/i2+2,3+2

InChI key

CURLTUGMZLYLDI-FTOQCNSHSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

Gas cylinder

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Press. Gas Liquefied gas

Storage Class Code

2A - Gases

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service