Skip to Content
Merck
All Photos(1)

Key Documents

578509

Sigma-Aldrich

Cyclopropylmethan-d2-ol

98 atom % D

Synonym(s):

a,a-Cyclopropanemethan-d2-ol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C3H5CD2OH
CAS Number:
Molecular Weight:
74.12
MDL number:
UNSPSC Code:
12352202
PubChem Substance ID:

Note: This product can be packaged on demand. For information on pricing, availability and packaging of custom sizes, please contact Stable Isotopes Customer Service

isotopic purity

98 atom % D

Assay

99% (CP)

bp

120 °C mmHg (lit.)

mass shift

M+2

SMILES string

[2H]C([2H])(O)C1CC1

InChI

1S/C4H8O/c5-3-4-1-2-4/h4-5H,1-3H2/i3D2

InChI key

GUDMZGLFZNLYEY-SMZGMGDZSA-N

Compare Similar Items

View Full Comparison

Show Differences

1 of 4

This Item
588121490652363049
mass shift

M+2

mass shift

M+2

mass shift

M+2

mass shift

M+2

isotopic purity

98 atom % D

isotopic purity

98 atom % D

isotopic purity

98 atom % D

isotopic purity

99 atom % D

Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

bp

120 °C mmHg (lit.)

bp

97 °C (lit.)

bp

141 °C (lit.)

bp

68 °C/15 mmHg (lit.)

Packaging

This product may be available from bulk stock and can be packaged on demand. For information on pricing, availability and packaging, please contact Stable Isotopes Customer Service.

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

95.0 °F - closed cup

Flash Point(C)

35 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service