578193
Poly[(methyl methacrylate)-co-(9-anthracenylmethyl methacrylate)]
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
composition
chromophore, 20 mol %
transition temp
Tg 103 °C (DSC)
fluorescence
λex 251 nm; λem 416 nm in chloroform
General description
Fluorescent polymer.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service